Skip to Content
Merck
All Photos(1)

Key Documents

69659

Sigma-Aldrich

pLysS DNA - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

1 μG
€174.00

€174.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 μG
€174.00

About This Item

UNSPSC Code:
41106602
NACRES:
NA.51

€174.00


Estimated to ship on28 May 2025


form

solid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

pLysS and pLysE are 4886bp plasmids constructed by insertion of the T7 lysozyme gene into the BamH I site of pACYC184 (1). These plasmids are not cloning vectors; they are used in λDE3 lysogenic hosts to suppress basal expression from the T7 promoter by producing T7 lysozyme, a natural inhibitor of T7 RNA polymerase. The two plasmids differ only by the orientation of the T7 lysozyme gene. In pLysS the T7 lysozyme coding sequence is in the antisense orientation relative to the tet promoter, so only a small amount of T7 lysozyme is produced. In pLysE large amounts of T7 lysozyme are produced from the tet promoter. The construct also contains the weak T7 φ3.8 promoter immediately following the lysozyme gene. The p15A origin of replication is compatible with those found in pBR322- and pUC-derived plasmids.

This product is covered under license from Brookhaven National Labs. Commercial entities need to obtain a research use license prior to purchase. Please contact your licensing department to confirm that your company already holds a research use license.

Warning

Toxicity: Standard Handling (A)

Other Notes

1. Studier , F.W. (1991) J. Mol. Biol. 219, 37-44.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service