Skip to Content
Merck
All Photos(1)

Key Documents

69390-M

Sigma-Aldrich

Bacteriophage CE6 - Novagen

λ phage for delivery of T7 RNA polymerase

Sign Into View Organizational & Contract Pricing

Select a Size

0.2 ML
€174.00

€174.00


Estimated to ship on30 May 2025



Select a Size

Change View
0.2 ML
€174.00

About This Item

UNSPSC Code:
41106602
NACRES:
NA.54

€174.00


Estimated to ship on30 May 2025


Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

λ phage for delivery of T7 RNA polymerase
Bacteriophage CE6 is a recombinant lambda phage containing the T7 RNA polymerase gene (Studier 1986). The polymerase gene is cloned into the int gene, such that it is transcribed from the pL and pI promoters of the phage during infection. λCE6 can be used to provide a source of T7 RNA polymerase to susceptible host cells carrying T7 expression plasmids. In general, it is used when a target gene is so toxic that the plasmid cannot be maintained in DE3 lysogenic hosts, or with alternative hosts that are not DE3 lysogens. The phage has λcI857 immunity and the Sam7 mutation, which requires supF hosts for lytic growth (neither lytic growth nor different immunity are required for T7 RNA polymerase synthesis). It is provided as a high titer lysate and includes host strains LE392 (rK12 mK12+) and ED8739 (rK12 mK12) as glycerol stocks. The 10-ml size is enough to induce up to 100 ml culture.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Components

•0.2 or 10 mlCE6 Lysate

•0.2 mlLE392 glycerol stock

•0.2 mlED8739 glycerol stock

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

Studier, F.W. and Moffatt, B.A. 1986 J. Mol. Biol. 189, 113.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is it necessary to include maltose?

    1 answer
    1. Yes, It is recommended to add maltose as it increases the lambda phage receptors on the surface of the E. coli cells, which is beneficial for the mutagenesis process.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service