Skip to Content
Merck
All Photos(1)

Key Documents

69025-M

Millipore

T7•Tag® Affinity Purification Kit

Immunoaffinity purification of T7•Tag fusion proteins

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€524.00

€524.00


Estimated to ship on29 March 2025



Select a Size

Change View
1 KIT
€524.00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

€524.00


Estimated to ship on29 March 2025


Quality Level

manufacturer/tradename

Novagen®

storage condition

do not freeze

storage temp.

2-8°C

General description

Immunoaffinity purification of T7•Tag fusion proteins
The T7•Tag Affinity Purification Kit is designed for rapid immunoaffinity purification of target proteins that carry the T7•Tag sequence (e.g., the 11 amino terminal aa of the T7 gene 10 protein). Purification is based on binding target proteins to T7•Tag Monoclonal Antibody that is covalently coupled to cross-linked agarose beads, washing away unbound proteins, and eluting at pH 2.2. Neutralization Buffer is included to limit protein exposure to low pH. Capacity will vary somewhat between different target proteins, but the beads are standardized to bind a minimum of 300 µg T7•Tag β-galactosidase per milliliter of settled resin. The beads can be used in either batch or column methods and can be regenerated a minimum of five times without loss of binding activity.

Components

•1 mlT7•Tag Antibody Agarose

•20 ml10X T7•Tag Bind/Wash Buffer

•20 ml10X T7•Tag Elution Buffer

•20 mlT7•Tag Neutralization Buffer

•1Chromatography Column

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
T7-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service