Skip to Content
Merck
All Photos(2)

Key Documents

681664

Sigma-Aldrich

Wnt Agonist I in DMSO

The Wnt Agonist I in DMSO, also referenced under CAS 853220-52-7, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Wnt Agonist I in DMSO, 2-Amino-4-(3,4-(methylenedioxy)benzylamino)-6-(3-methoxyphenyl)pyrimidine, Wnt Pathway Activator I in DMSO, Wnt Pathway Activator I in DMSO, 2-Amino-4-(3,4-(methylenedioxy)benzylamino)-6-(3-methoxyphenyl)pyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€114.00

€114.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
2 MG
€114.00

About This Item

Empirical Formula (Hill Notation):
C19H18N4O3
CAS Number:
Molecular Weight:
350.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€114.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

ambient

storage temp.

−70°C

SMILES string

NC1=NC(NCC2=CC=C(OCO3)C3=C2)=CC(C4=CC=CC(OC)=C4)=N1

InChI

1S/C19H18N4O3/c1-24-14-4-2-3-13(8-14)15-9-18(23-19(20)22-15)21-10-12-5-6-16-17(7-12)26-11-25-16/h2-9H,10-11H2,1H3,(H3,20,21,22,23)

InChI key

FABQUVYDAXWUQP-UHFFFAOYSA-N

General description

A cell-permeable, potent, and selective activator of Wnt signaling that does not inhibit the activity of GSK-3β (IC50 >60 µM). Shown to mimic the effect of Wnt and induce β-catenin and TCF (T-cell fate)-dependent transcriptional activity (EC50 = 700 nM in HEK-293T cells). Reported to cause substantial head defects during Xenopus embryonic development. The solid form of this compound (Cat. No. 681665) is also available.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (2 mg/228 µL) solution of Wnt Agonist I (Cat. No. 681665) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Liu, J., et al. 2005. Angew. Chem. Int. Ed.44, 1987.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service