Skip to Content
Merck
All Photos(1)

Key Documents

6805-OP

Millipore

Phenol:Chloroform:Isoamyl Alcohol

OmniPur® Grade, 25:24:1 ratio, phenol: >=99.0%, Suitable for DNA applications

Synonym(s):

phenol:Chloroform:Isoamyl Alcohol, 25:24:1

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
inquire
400 ML
€183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
inquire
400 ML
€183.00

About This Item

UNSPSC Code:
12190000
NACRES:
NA.07


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid (two-layer)

manufacturer/tradename

Calbiochem®

color

colorless to slightly yellow

shipped in

ambient

General description

Phenol saturated with TE buffer for DNA isolation. Comes at pH 6.7 for stability with included TE buffer to raise pH to 8.0 for DNA extraction.
pH 8.0±0.2: Gently mix contents of phenol: chloroform and buffer bottles. Allow layers to separate before use (2-4 h). pH 6.7±0.2: Use saturated phenol: chloroform as is without addition of buffer bottle. Solution is stable >1 year at 4°C.Phenol saturated with TE buffer for DNA isolation. Comes at pH 6.7 for stability with included TE buffer to raise pH to 8.0 for DNA extraction. Suitable for DNA applications. Molecular biology grade.

Specifications:
Assay [phenol]: ≥99.0%
pH of Saturated phenol at 25°C: 6.5-6.9
pH of TRIS-EDTA buffer: 7.8-8.2
UV Absorbance at 330 nm: ≤0.2AU
UV Absorbance at 405 nm: ≤0.1AU
UV Absorbance at 510 nm: ≤0.1AU





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Complete mitochondrial genome of critically endangered catfish Hemibagrus punctatus (Jerdon, 1849) and comparative analysis for insights into the phylogeny of hemibagrids through mitogenomic approach.: This study presents the complete mitochondrial genome of the critically endangered catfish Hemibagrus punctatus and offers comparative analyses for phylogenetic insights. Phenol:Chloroform:Isoamyl Alcohol was used in the extraction of high-quality DNA for genomic studies, facilitating the understanding of genetic diversity and conservation efforts (Chowdhury et al., 2024).

  • The changes in tissue histomorphology and quality of DNA in healthy human autopsied tissues stored at -20 °C and -150 °C.: This research examines histomorphological changes and DNA quality in human tissues stored at different temperatures. Phenol:Chloroform:Isoamyl Alcohol was employed for DNA extraction, critical for maintaining the integrity of genetic material in forensic and clinical applications (Radojevic et al., 2024).

  • The use of vitreous humour as a potential source of DNA for postmortem identification in forensic science.: This study explores the potential of vitreous humour as a DNA source for postmortem identification, utilizing Phenol:Chloroform:Isoamyl Alcohol for effective DNA extraction, enhancing forensic analysis accuracy (Čanović et al., 2024).

  • A new DNA extraction method (HV-CTAB-PCI) for amplification of nuclear markers from open ocean-retrieved faeces of an herbivorous marine mammal, the dugong.: This paper introduces an advanced DNA extraction method for nuclear markers from marine mammal feces. Phenol:Chloroform:Isoamyl Alcohol played a crucial role in optimizing DNA purity and yield, pivotal for genetic studies in marine biology (Ooi et al., 2023).

  • A comparison of five DNA extraction methods from degraded human skeletal remains.: This forensic study compares various DNA extraction methods for degraded skeletal remains. Phenol:Chloroform:Isoamyl Alcohol was highlighted for its effectiveness in recovering usable DNA, proving essential in forensic anthropology (Haarkötter et al., 2023).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service