Skip to Content
Merck
All Photos(1)

Key Documents

672112

Sigma-Aldrich

Urokinase, Human Urine

Synonym(s):

Urokinase, Human Urine, uPA

Sign Into View Organizational & Contract Pricing

Select a Size

10 KU
€324.00

€324.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 KU
€324.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€324.00


Check Cart for Availability

Request a Bulk Order

description

Merck USA index - 14, 9887

Quality Level

form

lyophilized solid

specific activity

≥100,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

storage temp.

−20°C

General description

Native urokinase from human urine. A thrombolytic agent that converts plasminogen to plasmin. A mixture of high molecular weight (54 kDa) and low molecular weight (33 kDa) urokinase.
Note: 1 unit = 1 CTA unit = 0.7 Ploug units. 1 KU = 1000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme equal to an international standard tested by the fibrinolytic method of Johnson, et al.

Physical form

Lyophilized from 200 mM mannitol, 100 mM NaCl, 50 mM Tris-HCl, 0.1% PEG, pH 7.5.

Preparation Note

Prepared from urine of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.
Reconstitute in distilled H₂O

Other Notes

Bell, W.R. 1975. Semin. Thromb. Res.2, 1.
Johnson, A.J., et al. 1969. Thromb. Diath. Haemorrh. 21, 259.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service