Skip to Content
Merck
All Photos(1)

Key Documents

655205

Millipore

TWEEN® 20

Non-ionic detergent

Synonym(s):

TWEEN® 20, Polysorbate 20, Polyoxyethylene (20) sorbitan monolaurate, PEG(20)sorbitan monolaurate, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
Inquire


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

TWEEN® 20, Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins.

Quality Level

description

RTECS - TR7400000

form

liquid

mol wt

~1228

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

refractive index

n20/D 1.468 (lit.)

CMC

59 μM (HLB number 16.7)

solubility

water: 10%

density

1.095 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

15-25°C

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-ionic detergent used for immunoprecipitation and for the solubilization of membrane-bound proteins (CMC: 59 µM; HLB number: 16.7).

Application

Non-ionic detergent

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

527.0 °F - Pensky-Martens closed cup

Flash Point(C)

275 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Wimmer et al.
Cell communication and signaling : CCS, 18(1), 61-61 (2020-04-12)
Cutaneous squamous cell carcinomas (cSCC) are the primary cause of premature deaths in patients suffering from the rare skin-fragility disorder recessive dystrophic epidermolysis bullosa (RDEB), which is in marked contrast to the rarely metastasizing nature of these carcinomas in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service