Skip to Content
Merck
All Photos(1)

Key Documents

65092E

Sigma-Aldrich

Decolorizer Gram Solution

HARLECO® Individual replacement for decolorizer component of the Gram Staining Set

Synonym(s):

Gram Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

form

liquid

mp

-88.88 °C

transition temp

flash point -20 °C

density

0.79 g/cm3

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

no temp limit

Application

HARLECO® Individual replacement for decolorizer component of the Gram Staining Set
Individual replacement for decolorizer component of the Gram Staining Set. Mixture of acetone and isopropanol used to decolorize Gram-negative bacteria.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service