Skip to Content
Merck
All Photos(1)

Key Documents

650211

Sigma-Aldrich

Trypsin, Excision Grade, Bovine Pancreas

Synonym(s):

Trypsin, Excision Grade, Bovine Pancreas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.

Assay

≥90% (HPLC)

Quality Level

form

solid

specific activity

≥80 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

trifluoroacetic acid: 0.01%
HCl: 1 mM

foreign activity

Chymotrypsin ≤0.05%

shipped in

ambient

storage temp.

2-8°C

General description

Native trypsin from bovine pancreas. Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis. Designed for protein sequencing or sequence verification, analysis of protein structural domains and cleavage of fusion proteins.
Native trypsin from bovine pancreas. Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis. Designed for protein sequencing or sequence verification, protein structural domains, and cleavage of fusion proteins.

Warning

Toxicity: Irritant (B)

Unit Definition

1 unit is the amount of enzyme that will convert 1 µmol of substrate per min at 37°C in 100 mM Tris pH 8.5

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Dilute into aqueous buffers (at neutral pH) just prior to use. Stock solutions are stable for up to 1 month at -20°C or for up to 1 week at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service