Skip to Content
Merck
All Photos(2)

Documents

648311

Millipore

Tris Base

≥99% dry basis (titration), crystalline solid, pH 10.3-11.3, Ultrol® Grade

Synonym(s):

Tris(hydroxymethyl)aminomethane, tris(Hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.25

product name

Tris Base, ULTROL® Grade, ≥99% dry basis (titration), useful in pH range 10.3-11.3

Quality Level

Assay

≥99% dry basis (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

water: 1 M

cation traces

heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

10-30°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Absorbance (1.0 M, H2O, 260 nm): <0.05.
Useful in the pH range of 10.3-11.3. Has a pKa of 8.1 at 25°C.
Widely used as a biological buffer (absorbance: <0.05 at 260 nm, 1.0 M, H2O). A 1.0 M solution in H2O has a pH range of 10.3-11.3 and a pKa of 8.3 at 20°C.

Application


  • Development of an Antigen Capture Lateral Flow Immunoassay for the Detection of Burkholderia pseudomallei.: This study uses Tris Base in the development of a lateral flow immunoassay for detecting Burkholderia pseudomallei, enhancing diagnostic capabilities in clinical microbiology (Nualnoi et al., 2024).

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aura E Ionescu et al.
International journal of molecular sciences, 20(24) (2019-12-19)
Eyes absent (EYA) are non-thiol-based protein tyrosine phosphatases (PTPs) that also have transcriptional co-activator functions. Their PTP activity is involved in various pathologies. Recently, we demonstrated that Src tyrosine kinase phosphorylates human EYA3 by controlling its subcellular localization. We also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service