Skip to Content
Merck
All Photos(1)

Key Documents

645900

Sigma-Aldrich

Triptolide, Tripterygium wilfordii

A novel diterpene triepoxide isolated from the Chinese herb Tripterygium wilfordii that acts as a potent immunosuppressant and anti-inflammatory agent.

Synonym(s):

Triptolide, Tripterygium wilfordii, PG490

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€171.00
5 MG
€686.00

€171.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€171.00
5 MG
€686.00

About This Item

Empirical Formula (Hill Notation):
C20H24O6
CAS Number:
Molecular Weight:
360.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€171.00


Estimated to ship on16 May 2025


Request a Bulk Order

Quality Level

description

RTECS - YK7751000

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@@]32[C@]5(O[C@H]5C[C@@H]6[C@@]3(CCC7=C6COC7=O)C)[C@@H]([C@]4(O[C@H]4[C@@H]21)C(C)C)O

InChI

1S/C20H24O6/c1-8(2)18-13(25-18)14-20(26-14)17(3)5-4-9-10(7-23-15(9)21)11(17)6-12-19(20,24-12)16(18)22/h8,11-14,16,22H,4-7H2,1-3H3/t11-,12-,13-,14-,16+,17-,18-,19+,20+/m0/s1

InChI key

DFBIRQPKNDILPW-CIVMWXNOSA-N

General description

A novel diterpene triepoxide isolated from the Chinese herb Tripterygium wilfordii that acts as a potent immunosuppressant and anti-inflammatory agent. Induces apoptosis in tumor cells by activating DEVD-cleaving caspases. Sensitizes tumor cells to TNF-α-induced apoptosis, and inhibits TNF-α-induced activation of NF-κB. Also inhibits T cell IL-2 expression at the level of purine-box/nuclear factor of activated T cells and NF-κB transcriptional activation. Also reported to inhibit mitogen activated protein kinase phosphatase-1 (MKP-1).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Activate DEVD-cleaving caspases
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Zhao, Q., et al. 2005. J. Biol. Chem.280, 8101.
Shepherd, E., et al. 2004. J. Biol. Chem.279, 54023.
Chan, M.A., et al. 1999. Phytother. Res. 13, 464.
Lee, K.Y., et al. 1999. J. Biol. Chem. 274, 13451.
Qiu, D., et al. 1999. J. Biol. Chem. 274, 13443.
Yang, Y., et al. 1998. Immunopharmacology40, 139.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ildikó Kristó et al.
Biochimica et biophysica acta. Molecular cell research, 1864(10), 1589-1604 (2017-05-31)
Current models imply that the evolutionarily conserved, actin-binding Ezrin-Radixin-Moesin (ERM) proteins perform their activities at the plasma membrane by anchoring membrane proteins to the cortical actin network. Here we show that beside its cytoplasmic functions, the single ERM protein of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service