Skip to Content
Merck
All Photos(1)

Key Documents

64571-M

Sigma-Aldrich

Wright-Giemsa Fuccillo Stain Modification

Wright-Giemsa Fuccillo Stain Modification

Harleco®, suitable for hematology

Synonym(s):

Hematology Stain, Hematology Solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco®

mp

-98 °C

density

0.791 g/cm3

λmax

520-530 nm (Eosin)
640-650 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology

hematology ((contains Methanol))

storage temp.

15-25°C

General description

HARLECO® Polychromatic stain for peripheral blood and bone marrow smears

Application

HARLECO® Polychromatic stain for peripheral blood and bone marrow smears
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Polychromatic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the mixture of stain to buffer for the Wright Giemsa Fucillo solution?

    1 answer
    1. As per the instructions located on page 1 of the Instructions for Use, the stain buffer mix is prepared by combining 50 ml of the Wright Giemsa Fucillo stain with 250 ml of buffer. Depending on user preference, the pH of the buffer should be either pH 6.4 or 6.8. It is conceivable that some users might prefer a slightly more alkaline pH buffer. The stain buffer mixture should remain effective for at least 3 hours, but it is possible that it might remain effective for as long as 24 hours or perhaps slightly longer. When the stain quality begins to deteriorate, the stain buffer mix should be replaced.

      Helpful?

Reviews

Active Filters

  1. Montreal, Qc
    • Reviews 2
    • Vote 1
    5 out of 5 stars.

    Beautiful stain

    We have great results with this stain for a manual technique. It works very well with the phosphate buffer with a pH of 6,4.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service