Skip to Content
Merck
All Photos(1)

Documents

6.18509

Sigma-Aldrich

Tetrabutylammonium hydroxide solution

10% in methanol

Synonym(s):

Tetrabutylammonium hydroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein:
3571228
MDL number:
UNSPSC Code:
12352119

form

solution

concentration

10% in methanol

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service