Skip to Content
Merck
All Photos(1)

Key Documents

616458

Sigma-Aldrich

TGF-β RI Kinase Inhibitor VII - CAS 666729-57-3 - Calbiochem

The TGF-β RI Kinase Inhibitor VII, also referenced under CAS 666729-57-3, controls the biological activity of TGF-β RI Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

TGF-β RI Kinase Inhibitor VII - CAS 666729-57-3 - Calbiochem, 1-(2-((6,7-Dimethoxy-4-quinolyl)oxy)-(4,5-dimethylphenyl)-1-ethanone, ALK5 Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 2 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable quinoline compound that acts a potent ATP-competitive inhibitor of ALK5/TGF-β Receptor I kinase activity (IC50 = 0.63 µM), while inhibiting c-Src only at higher concentrations (IC50 = 13 µM). Shown to block TGF-β-induced Smad2 phosphorylation and Smad2/3 transcription activity (IC50 = 0.5 and 0.37 µM, respectively) in human lung cancer epithelial cell line A549.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shimizu, T., et al. 2008. J. Med. Chem.51, 3326.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanan Ahmed et al.
Cancers, 12(8) (2020-08-01)
The signalling pathways involved in metastasis of oral adenoid cancer cells (TYS) in response to cancer-associated fibroblasts (COM D24) and normal oral mucosal fibroblasts (MM1) was examined. Metastatic cell behaviour was observed by cell-scatter, 3-D-collagen gel migration, and 3-D-spheroid invasion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service