Skip to Content
Merck
All Photos(1)

Key Documents

614350

Sigma-Aldrich

Tomatidine, HCl

A steroidal alkaloid that structurally resembles Cyclopamine, but lacks the capacity to inhibit Shh (Sonic Hedgehog) signaling. Reported to be non-teratogenic.

Synonym(s):

Tomatidine, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H45NO2 · xHCl
CAS Number:
Molecular Weight:
415.65 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥90% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMF: 2 mg/mL
DMSO: 2 mg/mL
ethanol: 2.5 mg/mL
chloroform: methanol (1:1): 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C27H45NO2.ClH/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4;/h16-24,28-29H,5-15H2,1-4H3;1H/t16-,17-,18-,19-,20+,21-,22-,23-,24-,25-,26-,27+;/m0./s1

InChI key

SXXHVPYRDFJKPG-XRWUXUKGSA-N

General description

A steroidal alkaloid that structurally resembles Cyclopamine (Cat. No. 239803), but lacks the capacity to inhibit Shh (Sonic Hedgehog) signaling. Reported to be non-teratogenic.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Negative control for Shh (Sonic Hedgehog) signaling
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required to achieve complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Berman, D.M., et al. 2003. Nature,425, 846.
Thayer, S.P., et al. 2003. Nature,425, 851.
Watkins, D.N., et al. 2003. Nature422, 313.
Cooper, M.K., et al. 1998. Science280, 1603.
Kim, S.K. and Melton, D.A. 1998. Proc. Natl. Acad. Sci. USA95, 13036.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service