Skip to Content
Merck
All Photos(1)

Key Documents

612080

Sigma-Aldrich

TIMP-1

≥90% (SDS-PAGE), liquid, MMP inhibitor, Calbiochem®

Synonym(s):

Tissue Inhibitor of Metalloproteinase-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€498.00

€498.00


Estimated to ship on20 May 2025



Select a Size

Change View
5 μG
€498.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€498.00


Estimated to ship on20 May 2025


Product Name

TIMP-1, Human Neutrophil Granulocyte, Native TIMP-1 from human granulocyte.

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native TIMP-1 from human granulocyte. Member of a family of inhibitors that participate in the activation and regulation of MMP activity. Forms a non-covalent stoichiometric complex with latent and active MMPs. Binds to pro-MMP-9 and MMP-9 via their C-terminal domains.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMP activity
Product does not compete with ATP.
Reversible: no

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0.05% Brij® 35 Detergent, 0.05% NaN₃, pH 7.0.

Preparation Note

Prepared from stimulated neutrophils that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kolkenbrock, H., et al. 1996. Biol. Chem. 377, 529.
Kolkenbrock, H., et al. 1995. Biol. Chem. Hoppe Seyler 376, 495.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service