Skip to Content
Merck
All Photos(1)

Key Documents

6110-OP

Millipore

MES Monohydrate

OmniPur® Grade, >=99.0%

Synonym(s):

OmniPur® MES, Monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
Inquire
1 KG
Inquire


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
Inquire
1 KG
Inquire

About This Item

Linear Formula:
(C4H8ON)CH2CH2SO3H · H2O
CAS Number:
MDL number:
UNSPSC Code:
12161704
NACRES:
NA.25


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

[S](=O)(=O)(O)CCN1CCOCC1

InChI

1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)

InChI key

SXGZJKUKBWWHRA-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
pKa at 25°C: 5.50-6.50
Purity: ≥99.0%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service