Skip to Content
Merck
All Photos(1)

Key Documents

605225

Sigma-Aldrich

Thrombospondin, Human Platelets

Synonym(s):

Thrombospondin protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€324.00

€324.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€324.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€324.00


Estimated to ship on16 May 2025


Request a Bulk Order

description

Merck USA index - 14 9387

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

wet ice

storage temp.

−70°C

General description

Native thrombospondin purified from human platelets. Adhesive glycoprotein released in response to platelet activation by α-thrombin. An angiogenesis inhibitor that blocks the FGF-induced neovascularization of the rat cornea. An inducer of platelet aggregation. Synthesized by a number of different cells and secreted into the extracellular matrix. Involved in the regulation of cellular proliferation.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 600 mM NaCl, 20 mM Tris, 1 mM CaCl₂, 20% sucrose, pH 7.8.

Preparation Note

Please refer to lot-specific reconstitution volume on the vial label.
Prepared from platelets that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for 1 week at 4°C or for at least 1 month at -70°C. Product is extremely adherent to surfaces; keep frozen (-70°C) before use and use siliconized surfaces to insure product recovery.

Other Notes

Armstrong, L.C., et al. 2002. Mol. Biol. Cell13, 1893.
Michaund, M. and Poyet, O. 1994. Anticancer Res. 14, 1127.
Castle, V.P., et al. 1993. J. Biol. Chem. 268, 2899.
Koch, A.E., et al. 1993. Pathobiology 61, 1.
Tolsma, S.S., et al. 1993. J. Cell Biol.122, 497.
Tuszynski, G.P., et al. 1993. J. Cell Biol. 120, 513.
Schon, P., et al. 1992. Eur. J. Cell Biol. 59, 329.
Good, D.J., et al. 1990. Proc. Natl. Acad. Sci. USA87, 6624.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service