Skip to Content
Merck
All Photos(1)

Key Documents

575545

Sigma-Aldrich

XAV939

≥97% (HPLC), solid, Tankyrase1/2 inhibitor, Calbiochem

Synonym(s):

Tankyrase1/2 Inhibitor, XAV939, 2-(4-(Trifluoromethyl)phenyl)-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol, Wnt Pathway Inhibitor V, TNKS1/2 Inhibitor I

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€221.00

€221.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€221.00

About This Item

Empirical Formula (Hill Notation):
C14H11F3N2OS
Molecular Weight:
312.31
UNSPSC Code:
12352200
NACRES:
NA.54

€221.00


Estimated to ship on19 May 2025


Request a Bulk Order

Product Name

Tankyrase1/2 Inhibitor, XAV939, The Tankyrase1/2 Inhibitor, XAV939 controls the biological activity of Tankyrase1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable dihydrothiopyranopyrimidinol that binds TNKS1/PARP5a and TNKS2/PARP5b with high affinity (Kd = 99 and 93 nM, respectively) and potently inhibits their PARsylation (poly ADP-ribosylation) activity (IC50 = 11 and 4 nM, respectively), while exhibiting much lower activity against PARP1 and PARP2 (IC50 = 2.194 and 0.114 µM, respectively). TNKS, but not PARP1/2, function knockdown by siRNA or XAV939 treatment suppresses cellular axin1/2 PARsylation and ubiquitination/proteasomal degradation, resulting in axin build-up, β-catenin destruction, and blockage of Wnt signaling. XAV939 is shown to inhibit the growth of β-catenin-dependent DLD-1, but not that of β-catenin-independent RKO, cells (by >95% vs. no effect at 3.3 µM, respectively) in vitro and prevent the regeneration of zebrafish tail fin (by ~70% at 7 d post-amputation, 5 µM) after surgical removal in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Huang, S.M., et al. 2009. Nature461, 614.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Pedone et al.
iScience, 25(2), 103756-103756 (2022-02-08)
The Wnt/β-catenin pathway is involved in development, cancer, and embryonic stem cell (ESC) maintenance; its dual role in stem cell self-renewal and differentiation is still controversial. Here, by applying an in vitro system enabling inducible gene expression control, we report that
Bat-Erdene Jargalsaikhan et al.
Viruses, 16(6) (2024-06-27)
A gene delivery system utilizing lentiviral vectors (LVs) requires high transduction efficiency for successful application in human gene therapy. Pseudotyping allows viral tropism to be expanded, widening the usage of LVs. While vesicular stomatitis virus G (VSV-G) single-pseudotyped LVs are
Hirosato Ideno et al.
iScience, 25(10), 105140-105140 (2022-10-04)
Various culture methods have been developed for maintaining human pluripotent stem cells (PSCs). These PSC maintenance methods exhibit biased differentiation; for example, feeder-dependent PSCs efficiently yield cerebral organoids, but it is difficult to generate organoids from feeder-free PSCs. It remains
Induction of fetal meiotic oocytes from embryonic stem cells in cynomolgus monkeys.
Gyobu-Motani, et al.
The Embo Journal, 42, e112962-e112962 (2023)
Masashi Hada et al.
Genes & development, 36(1-2), 84-102 (2022-01-08)
The placenta is a highly evolved, specialized organ in mammals. It differs from other organs in that it functions only for fetal maintenance during gestation. Therefore, there must be intrinsic mechanisms that guarantee its unique functions. To address this question

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service