Skip to Content
Merck
All Photos(1)

Key Documents

575310

Sigma-Aldrich

T0901317

A cell-permeable, nonsterol, benzenesulfonamide compound that acts as a highly selective and potent liver X receptor agonist (EC₅₀ of 20 nM for LXRα).

Synonym(s):

T0901317, TO-901317, N-(2,2,2-Trifluoroethyl)-N-[4-(2,2,2-trifluoro-1-hydroxy-1-trifluoromethylethyl)phenyl]sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€230.00

€230.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C17H12F9NO3S
CAS Number:
Molecular Weight:
481.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€230.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 50 mg/mL
ethanol: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

FC(F)(F)CN([S](=O)(=O)c2ccccc2)c1ccc(cc1)C(O)(C(F)(F)F)C(F)(F)F

InChI

1S/C17H12F9NO3S/c18-14(19,20)10-27(31(29,30)13-4-2-1-3-5-13)12-8-6-11(7-9-12)15(28,16(21,22)23)17(24,25)26/h1-9,28H,10H2

InChI key

SGIWFELWJPNFDH-UHFFFAOYSA-N

General description

A cell-permeable, nonsterol, benzenesulfonamide compound that acts as a highly selective and potent liver X receptor agonist (EC50 of 20 nM for LXRα). Reported to induce the expression of genes associated with fatty acid biosynthesis and raise both the levels of serum HDL cholesterol and triglyceride in mice. Also reduces the development of atherosclerosis in LDL receptor-deficient mice and lowers plasma glucose in diabetic rodents.

Biochem/physiol Actions

Cell permeable: yes
EC50 = 20 nM for LXRα
Primary Target
Liver X receptor (LXRα)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Terasaka, N., et al. 2003. FEBS Lett.536, 6.
Cao, G., et al. 2002. J. Biol. Chem.278, 1131.
Field, F.J., et al. 2002. Biochem. J.368, 855.
Fukumoto, H., et al. 2002. J. Biol. Chem.277, 48508.
DeBose-Boyd, R.A., et al. 2001. Proc. Natl. Acad. Sci. USA98, 1477.
Repa, J.J., et al. 2000. Science289, 1524.
Schultz, J.R., et al. 2000. Genes Dev.14, 2831.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service