Skip to Content
Merck
All Photos(1)

Key Documents

574776

Sigma-Aldrich

Swinholide A, Theonella swinhoei

A dimeric dilactone macrolide that is cytotoxic to a variety of cancer cell lines in vitro.

Synonym(s):

Swinholide A, Theonella swinhoei

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€267.00

€267.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€267.00

About This Item

Empirical Formula (Hill Notation):
C78H132O20
CAS Number:
Molecular Weight:
1389.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€267.00


Estimated to ship on10 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

acetone: soluble
ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@H](C[C@@H](C[C@@H]1C)OC)CC[C@@H]([C@H](O)[C@@H]([C@H]2OC(=O)\C=C\C(=C\C[C@@H](C[C@H]3O[C@H](C[C@@H]([C@H]([C@H](C[C@H]([C@@H]([C@H](OC(=O)\C=C\C(=C\C[C@@H](C[C@H]5O[C@H](C[C@@H]([C@H]([C@H](C[C@H]([C@@H]2C)O)O)C)OC)CC=C5)O)\C)[C@H]([C@@H](O)[C@H](CC[

InChI

1S/C78H132O20/c1-45-23-29-57(79)37-59-19-17-21-61(95-59)41-71(91-15)52(8)68(82)44-70(84)54(10)78(56(12)76(88)48(4)28-32-64-40-66(90-14)36-50(6)94-64)98-74(86)34-26-46(2)24-30-58(80)38-60-20-18-22-62(96-60)42-72(92-16)51(7)67(81)43-69(83)53(9)77(97-73(85)3

InChI key

RJVBVECTCMRNFG-ANKJNSLFSA-N

General description

A dimeric dilactone macrolide isolated from the marine sponge Theonella swinhoei. Cytotoxic to a variety of cancer cell lines in vitro. Disrupts the actin cytoskeleton by sequestering actin dimers with a binding stoichiometry of 1:1, and rapidly severing F-actin. Also possesses antifungal activity.
A dimeric dilactone macrolide that is cytotoxic to a variety of cancer cell lines in vitro. Disrupts the actin cytoskeleton by sequestering actin dimers with a binding stoichiometry of 1:1, and rapidly severing F-actin. Also possesses antifungal properties.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Disrupts the actin cytoskeleton
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Saito, S.Y., et al. 1998. J. Biochem. (Tokyo) 123, 571.
Bubb, M.R., et al. 1995. J. Biol. Chem. 270, 3463.
Kobayashi, M., et al. 1994. Chem. Pharm. Bull. 42, 19.
Doi, M., et al. 1991. J. Org. Chem. 56, 3629.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service