Skip to Content
Merck
All Photos(1)

Key Documents

574713

Sigma-Aldrich

Syk Inhibitor III

The Syk Inhibitor III, also referenced under CAS 1485-00-3, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Syk Inhibitor III, 3,4-Methylenedioxy-β-nitrostyrene

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€139.00

€139.00


Estimated to ship on29 May 2025



Select a Size

Change View
50 MG
€139.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO4
CAS Number:
Molecular Weight:
193.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€139.00


Estimated to ship on29 May 2025


description

RTECS - WL5270000

Quality Level

Assay

≥98% (GC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to brown

solubility

DMSO: 25 mg/mL (Use only fresh DMSO.)

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])\C=C/c1cc2c(cc1)OCO2

InChI

1S/C9H7NO4/c11-10(12)4-3-7-1-2-8-9(5-7)14-6-13-8/h1-5H,6H2/b4-3-

InChI key

KFLWBZPSJQPRDD-ARJAWSKDSA-N

General description

A cell-permeable nitrostyrene compound that acts as a selective inhibitor of Syk kinase activity (IC50 = 2.5 µM). It inhibits Src only at much higher concentrations (IC50 = 29.3 µM) and exhibits little effect against Fak and JAK2. Shown to potently inhibit platelet aggregation induced by various stimulators, including U46619, ADP, arachidonic acid, collagen, thrombin, ionophore A23187, and PDBu (IC50 ranges from 2.1 to 25.9 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Syk
Product does not compete with ATP.
Reversible: no
Target IC50: 2.5 µM against Syk kinase activity

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, W.Y., et al. 2006. Mol. Pharmacol.70, 1380.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service