Skip to Content
Merck
All Photos(1)

Key Documents

573121

Sigma-Aldrich

SMER28

A cell-permeable quinazoline compound that augments the cytostatic effects of Rapamycin and acts as an autophagy stimulator both in mammalian cultures in vitro and in a Drosophila melanogaster HD model in vivo.

Synonym(s):

SMER28, (6-Bromo-4-allylamino)quinazoline, Small- Molecule Enhancer of Rapamycin-28, Autophagy Inducer II, (6-Bromo-4-allylamino)quinazoline, Small-Molecule Enhancer of Rapamycin-28, Autophagy Inducer II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€171.00

€171.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€171.00

About This Item

Empirical Formula (Hill Notation):
C11H10BrN3
CAS Number:
Molecular Weight:
264.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€171.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

ethanol: 10 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1cc2c(ncnc2NCC=C)cc1

InChI

1S/C11H10BrN3/c1-2-5-13-11-9-6-8(12)3-4-10(9)14-7-15-11/h2-4,6-7H,1,5H2,(H,13,14,15)

InChI key

BCPOLXUSCUFDGE-UHFFFAOYSA-N

General description

A cell-permeable quinazoline compound that augments the cytostatic effects of Rapamycin (Cat. Nos. 553210 and 553211) in Saccharomyces cerevisiae and acts as an autophagy stimulator both in mammalian cultures in vitro and in a Drosophila melanogaster HD model in vivo. Although SMER28 and Rapamycin do exhibit additive effects on the clearance of cellular autophagy substrates, SMER28 functions independently of Rapamycin by presumably acting on celluar target(s) downstream of the Rapamycin/FKBP12 target, mTOR.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Tian, Y., et al. 2011. FASEB J.25, 1934.Sarkar, S., et al. 2007. Nat. Chem. Biol.3, 331.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service