Skip to Content
Merck
All Photos(1)

Key Documents

573100

Millipore

SATA

Synonym(s):

SATA, N-Succinimidyl-S-acetylthioacetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€184.00

€184.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 MG
€184.00

About This Item

Empirical Formula (Hill Notation):
C8H9NO5S
CAS Number:
Molecular Weight:
231.23
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.47

€184.00


Estimated to ship on26 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CC(=O)ON1C(=O)CCC1=O)C(=O)C

InChI

1S/C8H9NO5S/c1-5(10)15-4-8(13)14-9-6(11)2-3-7(9)12/h2-4H2,1H3

InChI key

FLCQLSRLQIPNLM-UHFFFAOYSA-N

General description

A useful agent for introducing thiol groups, in protected form, into various proteins. Reacts with primary amines to add protected sulfhydryl groups. When treated with hydroxylamine, yields a free sulfhydryl group that can be conjugated to maleimide-modified proteins.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Duncan, R.J., et al. 1983. Anal. Biochem.132, 68.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service