Skip to Content
Merck
All Photos(1)

Key Documents

573098

Sigma-Aldrich

JAK2 Inhibitor III, SD-1029

The JAK2 Inhibitor III, SD-1029, also referenced under CAS 118372-34-2, controls the biological activity of JAK2.

Synonym(s):

JAK2 Inhibitor III, SD-1029, 9-(3,4-Dichlorophenyl)-2,7-bis(dimethylaminomethyl)-3,4,5,6,7,9-hexahydro-2H-xanthene-1,8-dione, 2HBr, NSC 371488

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€205.00

€205.00


Estimated to ship on18 April 2025



Select a Size

Change View
10 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C25H30Cl2N2O3 · 2HBr
CAS Number:
Molecular Weight:
639.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on18 April 2025


Quality Level

Assay

≥80% (sum of isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable xanthenedione compound that acts as a JAK2-selective inhibitor. It exhibits much less effect on the cellular phosphorylation levels of Jak1 and Src and shows no significant inhibition towards a panel of 49 other protein kinases in cell-free kinase assays at a concentration of 10 µM. Effectively inhibits cell proliferation in JAK-activated BA/F3 cell lines as well as phosphorylation and nuclear translocation of cellular Stat3.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Duan, Z., et al. 2006. Clin. Cancer Res.12, 6844.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service