Skip to Content
Merck
All Photos(1)

Key Documents

5677

Sigma-Aldrich

Spermine, Tetrahydrochloride

Synonym(s):

Spermine, Tetrahydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€336.00

€336.00


Estimated to ship on22 May 2025



Select a Size

Change View
5 G
€336.00

About This Item

Empirical Formula (Hill Notation):
C10H26N4 · 4HCl
CAS Number:
Molecular Weight:
348.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€336.00


Estimated to ship on22 May 2025


description

RTECS - EJ7230000
Merck USA index - 14, 8743

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Cl-].[Cl-].[Cl-].[Cl-].[N+H2](CCC[N+H3])CCCC[N+H2]CCC[N+H3]

InChI

1S/C10H26N4.4ClH/c11-5-3-9-13-7-1-2-8-14-10-4-6-12;;;;/h13-14H,1-12H2;4*1H

InChI key

XLDKUDAXZWHPFH-UHFFFAOYSA-N

General description

Polyamine that plays an important role in the regulation of cellular proliferation and differentiation. Acts as an inhibitor of phopholipase C-α and an activator of phopholipase C-δ. Has both stimulatory and inhibitory effects on NMDA receptors depending on the subunit composition of the receptor. Inhibits neuronal NOS. Inhibits DNA fragmentation and morphological apoptosis.

Biochem/physiol Actions

Primary Target
phospholipase C α
Secondary Target
nNOS

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, purge with inert gas, and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C under inert gas.

Other Notes

Igarashi, K. and Williams, K. 1995. J. Pharmacol. Exp. Ther. 272, 1101.
Escargueil-Blanc, I., et al. 1994. FASEB J.8, 1075.
Hu, J., et al. 1994. Neurosci. Lett. 175, 41.
Tompkins, T.A. and Moscarello, M.A. 1994. Biochim. Biophys. Acta1206, 1208.
Trubiani, O., et al. 1994. Biochem. Mol. Biol. Int.32, 1085.
Svensson, F., et al. 1993. Mol. Cell. Biochem.124, 141.
Ask, A., et al. 1992. Cancer Lett.66, 29.
Chida, N., et al. 1992. Brain Res.593, 57.
Khan, A.U., et al. 1992. Proc. Natl. Acad. Sci. USA89, 11428.
Messena, L., et al. 1992. J. Leukoc. Biol. 52, 585.
Pawelczyk, T., et al. 1992. Arch. Biochem. Biophys.297, 328.
Shantz, L.M., et al. 1992. Biochem. J.288, 511.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanislau Yatskevich et al.
Cell reports, 34(13), 108929-108929 (2021-04-01)
The anaphase-promoting complex/cyclosome (APC/C) is an E3 ubiquitin ligase that controls cell cycle transitions. Its regulation by the spindle assembly checkpoint (SAC) is coordinated with the attachment of sister chromatids to the mitotic spindle. APC/C SUMOylation on APC4 ensures timely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service