Skip to Content
Merck
All Photos(1)

Key Documents

567570

Sigma-Aldrich

Spectinomycin, Dihydrochloride, Pentahydrate, Streptomyces sp.

A broad spectrum aminoglycoside antibiotic that contains two glucose moieties. Footprint studies indicate that spectinomycin exerts regional effects on ribosomal structure.

Synonym(s):

Spectinomycin, Dihydrochloride, Pentahydrate, Streptomyces sp.

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€235.00

€235.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 G
€235.00

About This Item

Empirical Formula (Hill Notation):
C14H24N2O7 · 2HCl · 5H2O
CAS Number:
Molecular Weight:
495.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€235.00


Estimated to ship on29 May 2025


Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[N+H2]([C@@H]1[C@H]2O[C@@]3([C@H](O[C@@H]2[C@H]([C@@H]([C@@H]1O)[N+H2]C)O)O[C@@H](CC3=O)C)O)C.O.O.O.O.O

InChI

1S/C14H24N2O7.2ClH.5H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;;;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H;5*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;;;/m1......./s1

InChI key

DCHJOVNPPSBWHK-UXXUFHFZSA-N

General description

A broad spectrum aminoglycoside antibiotic that contains two glucose moieties. Footprint studies indicate that spectinomycin exerts regional effects on ribosomal structure. A resistance gene (aph) which encodes a spectinomycin has been cloned from Legionella pneumophila (disease agent for Legionnaire’s disease) kinase and expressed in E. coli. Potency: ≥500 µg/mg.

Biochem/physiol Actions

Primary Target
Footprint studies indicate that spectinomycin exerts regional effects on ribosomal structure

Warning

Toxicity: Standard Handling (A)

Other Notes

Thompson, P.R., et al. 1998. J. Biol. Chem. 273, 14788.
Noah, J., and Wollenzien, P. 1997. Nucleic Acids Symp. Ser. 36, 178.
Suter, T.M., et al. 1997. Antimicrob. Agents Chemother. 41, 1385.
Thom, G., and Prescott, C.D. 1997. Bioorg. Med. Chem. 5, 1081.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service