Skip to Content
Merck
All Photos(1)

Key Documents

567370

Sigma-Aldrich

Smac-N7 Peptide

A peptide that contains the N-terminal seven residues of Smac (Second Mitochondria-derived Activator of Caspases, also known as DIABLO) and promotes procaspase-3 activation at around 10 µM.

Synonym(s):

Smac-N7 Peptide, H-AVPIAQK-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H59N9O9
Molecular Weight:
725.88
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A peptide that contains the N-terminal seven residues of Smac (Second Mitochondria-derived Activator of Caspases, also known as DIABLO) and promotes procaspase-3 activation at around 10 µM. Reported to bind to the BIR2 and BIR3 (baculovirus IAP repeat) domains of XIAP (X-linked inhibitor of apoptosis proteins) with a Kd ~800 nM. Promotes the catalytic activity of caspase-9 in the presence of XIAP, suggesting that Smac competes with caspase-9 for binding to the same pocket on the surface of XIAP.

Biochem/physiol Actions

Cell permeable: no
Kd ~800 nM in binding to the BIR2 and BIR3 (baculovirus IAP repeat) domains of XIAP (X-linked inhibitor of apoptosis proteins)
Primary Target
Promotes the catalytic activity of caspase-9 in the presence of XIAP
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ala-Val-Pro-Ile-Ala-Gln-Lys-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Srinivasula, S.M., et al. 2001. Nature410, 112.
Chai, J., et al. 2000. Nature406, 855.
Liu, Z., et al. 2000. Nature408, 1004.
Srinivasula, S.M., et al. 2000. J. Biol. Chem.275, 36152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service