Skip to Content
Merck
All Photos(1)

Documents

565792-M

Sigma-Aldrich

γ-Secretase Inhibitor XXIII, Aβ42-Selective

The γ-Secretase Inhibitor XXIII, Aβ42-Selective controls the biological activity of γ-Secretase.

Synonym(s):

γ-Secretase Inhibitor XXIII, Aβ42-Selective

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H39N7O
Molecular Weight:
513.68
UNSPSC Code:
12352203

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

General description

A cell-permeable purine compound that acts as a highly potent and selective inhibitor against γ-secretase-mediated production of Aβ42 over Aβ40 (IC50 = 21 and 350 nM, respectively) and Notch1 (Notch IC50/ Aβ42 IC50 >2400) in cultures in vitro. Reported to be blood-brain barrier-permeant and orally bioavailable in APP-YAC transgenic mice and effectively reduce Aβ42 (by 73%), but not Aβ40, level in brain (100 mg/kg, p.o.) in vivo.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Rivkin, A., et al. 2010. Bioorg. Med. Chem. Lett.20, 2279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service