Skip to Content
Merck
All Photos(1)

Key Documents

565764

Sigma-Aldrich

γ-Secretase Substrate, Fluorogenic

An internally quenched fluorogenic peptide substrate containing the C-terminal β-APP (amyloid β-peptide precursor protein) amino acid sequence that is cleaved by γ-secretase.

Synonym(s):

γ-Secretase Substrate, Fluorogenic, NMA-GGVVIATVK(DNP)- DR DR DR-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€214.00

€214.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C70H116N26O18
Molecular Weight:
1609.83
UNSPSC Code:
12352202
NACRES:
NA.54

€214.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 5 mg/mL
5% acetic acid: soluble

shipped in

ambient

storage temp.

−20°C

General description

An internally quenched fluorogenic peptide substrate containing the C-terminal β-APP (amyloid β-peptide precursor protein) amino acid sequence that is cleaved by γ-secretase. Shown to be sensitive and useful for assaying γ-secretase activity. The proteolysis at the Aβ40-, Aβ42-, and Aβ43-generating cleavage sites results in enhanced fluorescence. Excitation max.: ~355 nm; emission max.: ~440 nm.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

NMA-Gly-Gly-Val-Val-Ile-Ala-Thr-Val-Lys(DNP)-D-Arg-D-Arg-D-Arg-NH₂

Reconstitution

Following reconstitution, aliquot, flush with a stream of inert gas, and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Farmery, M.R., et al. 2003. J. Biol. Chem.278, 24277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service