Skip to Content
Merck
All Photos(1)

Key Documents

561300

Sigma-Aldrich

E-Selectin, Human, Recombinant, CHO Cells

Synonym(s):

CD62E, ELAM-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€587.00

€587.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€587.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€587.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1 EU/μg Endotoxin (EU/μg of E-Selectin)

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human E-selectin consisting of 535 amino acids minus the transmembrane and cytoplasmic domains expressed in CHO cells. This glycoprotein is transiently expressed on vascular endothelial cell surfaces where it is able to bind to ligands such as SLex found on leukocytes. This initial binding event is followed by a second interaction involving ICAM and VCAM-1 that leads to vascular penetration and leukocyte invasion into the extracellular matrix tissue.

Biochem/physiol Actions

Shown to bind to E-selectin ligands on U937 cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from Dulbecco′s PBS with stabilizer.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Reconstitute to 1 mg/ml with sterile H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service