Skip to Content
Merck
All Photos(1)

Key Documents

555580

Millipore

Ribavirin

A non-interferon-inducing, broad spectrum antiviral nucleoside. Reported to be active against RSV, HIV, HCV, and other infectious agents. Its metabolite, ribavirin-5′-phosphate, inhibits inosine monophosphate (IMP) dehydrogenase.

Synonym(s):

Ribavirin, 1-β-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide, Virazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
41116107

Quality Level

Assay

≥99% (HPLC)
≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 20 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[n]2(nc(nc2)C(=O)N)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C8H12N4O5/c9-6(16)7-10-2-12(11-7)8-5(15)4(14)3(1-13)17-8/h2-5,8,13-15H,1H2,(H2,9,16)/t3-,4-,5-,8-/m1/s1

InChI key

IWUCXVSUMQZMFG-AFCXAGJDSA-N

General description

A non-interferon-inducing, broad spectrum antiviral nucleoside that is active against RSV, HIV, HCV and other infectious agents. Inhibits the growth of human myeloma 8226 cells (IC50 = 40 µM) and ovarian carcinoma OVCAR-5 cells (IC50 = 35 µM). Its metabolite, ribavirin-5′-phosphate, inhibits inosine monophosphate (IMP) dehydrogenase. Antiviral effects of Ribavirin can be reversed by depriving infected cells of methionine or isoleucine.

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Other Notes

Li, W., et al. 1999. Oncol. Res.11, 243.
Honda, Y., et al. 1994. Antimicrob. Agents Chemother.38, 653.
Liao, H.J. and Stollar, V. 1993. Antiviral Res. 22, 285.
Kakumu, S., et al. 1993. Gastroenterology105, 507.
Sidwell, R.W., et al. 1979. Pharmacol. Ther.6, 123.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service