Skip to Content
Merck
All Photos(1)

Key Documents

551600-M

Millipore

Quercetin, Dihydrate

A cell-permeable and reversible inhibitor of PIM1 kinase (IC₅₀ = 43 nM), PI 3-K (IC₅₀ = 3.8 µM) and phospholipase A2 (IC₅₀ = 2 µM).

Synonym(s):

Quercetin, Dihydrate, 3,3ʹ,4ʹ,5,7-Pentahydroxyflavone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€57.40

€57.40


Estimated to ship on22 March 2025



Select a Size

Change View
100 MG
€57.40

About This Item

Empirical Formula (Hill Notation):
C15H10O7 · 2H2O
CAS Number:
Molecular Weight:
338.27
MDL number:
UNSPSC Code:
12352200

€57.40


Estimated to ship on22 March 2025


Quality Level

Assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

acetic acid: 1 mg/mL
aqueous base: 1 mg/mL
DMSO: 5 mg/mL

storage temp.

10-30°C

SMILES string

[o]1c2c([c](c(c1c3cc(c(cc3)O)O)O)=O)c(cc(c2)O)O.O.O

InChI

1S/C15H10O7.2H2O/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6;;/h1-5,16-19,21H;2*1H2

InChI key

GMGIWEZSKCNYSW-UHFFFAOYSA-N

General description

A cell-permeable and reversible inhibitor of PIM1 kinase (IC50 = 43 nM), PI 3-K (IC50 = 3.8 µM) and phospholipase A2 (IC50 = 2 µM). Also inhibits mitochondrial ATPase, phosphodiesterases, and protein kinase C. Induces apoptosis in K562, Molt-4, Raji, and MCAS tumor cell lines. Reported to activate sirtuins and promote the survival of eukaryotic cells.
A cell-permeable inhibitor of PIM1 kinase (IC50 = 43 nM), PI 3-K (IC50 = 3.8 µM) and phospholipase A2 (IC50 = 2 µM). Also inhibits mitochondrial ATPase, phosphodiesterases, and protein kinase C. Induces apoptosis in K562, Molt-4, Raji, and MCAS tumor cell lines. Reported to activate sirtuins and promote the survival of eukaryotic cells.

Biochem/physiol Actions

Primary Target
PI 3-kinase
Target IC50: 43 nM, 3.8 µM, 2 µM against PIM1 kinase, PI 3-K, and phospholipase A2, respectively

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Other Notes

Bullock, A.N., et al. 2005. J. Med. Chem.48, 7604.
Howitz, K.T., et al. 2003. Nature425, 191.
Wei, Y.Q., et al. 1994. Cancer Res.54, 4952.
Shibasaki, F., et al. 1993. Biochem. J.289, 227.
Matter, W.F., et al. 1992. Biochem. Biophys. Res. Comm.186, 624.
Levy, J., et al. 1984. Biochem. Biophys. Res. Commun.123, 1227.
Graziani, Y., et al. 1983. Eur. J. Biochem.135, 583.
Gschwendt, M., et al. 1983. Biochem. Biophys. Res. Commun. 117, 444.
Ruckstuhl, M., et al. 1979. Biochem. Pharmacol.28, 535.
Beretz, A., et al. 1978. Experientia 34, 1054.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service