Skip to Content
Merck
All Photos(1)

Key Documents

540500

Sigma-Aldrich

Purvalanol A

A potent, cell-permeable, reversible, and selective inhibitor of cyclin-dependent kinases.

Synonym(s):

Purvalanol A, 2-(1R-Isopropyl-2-hydroxyethylamino)-6-(3-chloroanilino)-9-isopropyl-purine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€145.00

€145.00


Estimated to ship on19 May 2025



Select a Size

Change View
1 MG
€145.00

About This Item

Empirical Formula (Hill Notation):
C19H25ClN6O
CAS Number:
Molecular Weight:
388.89
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€145.00


Estimated to ship on19 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1cc(ccc1)Nc2nc(nc3[n](cnc32)C(C)C)N[C@@H](CO)C(C)C

InChI

1S/C19H25ClN6O/c1-11(2)15(9-27)23-19-24-17(22-14-7-5-6-13(20)8-14)16-18(25-19)26(10-21-16)12(3)4/h5-8,10-12,15,27H,9H2,1-4H3,(H2,22,23,24,25)/t15-/m0/s1

InChI key

PMXCMJLOPOFPBT-HNNXBMFYSA-N

General description

A potent, cell-permeable, reversible, and selective inhibitor of cyclin-dependent kinases (Cdks; IC50 = 4 nM for Cdc2-cyclin B; IC50 = 70 nM for Cdk2-cyclin A; IC50 = 35 nM for Cdk2-cyclin E; and IC50 = 75 nM for Cdk5-p35).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cdc2/cyclin B
Product does not compete with ATP.
Reversible: yes
Target IC50: 4 nM for Cdc2/cyclin B; 70 nM for Cdk2/cyclin A; 35 nM for Cdk2/cyclin E; 75 nM for Cdk5/p35

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Gray, N.S., et al. 1998. Science 281, 533.

Legal Information

Sold under license of U.S. Patents 6,617,331 and 6,803,371.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service