Skip to Content
Merck
All Photos(1)

Key Documents

540223

Sigma-Aldrich

Purmorphamine

≥98% (HPLC), liquid, Hedgehog signaling activator, Calbiochem

Synonym(s):

InSolution Purmorphamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€199.00

€199.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€199.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.77

€199.00


Estimated to ship on26 April 2025


Request a Bulk Order

Product Name

Purmorphamine, InSolution, ≥98%, 50 mM in DMSO, induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

storage temp.

−20°C

General description

A cell-permeable purine compound that induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells (EC50 = 1 µM) and in MC3T3-E1 lineage-committed preosteoblasts. Also shown to induce transdifferentiation in pre-adipocytes and myoblasts. Its effects are reported to be synergistic with bone morphogenetic protein-4 (BMP-4). Reported to induce osteogenesis by activation of the Hedgehog signaling pathway.
Purmorphamine is a cell-permeable purine compound that induces osteoblast differentiation in C3H10T1/2 multipotent mesenchymal progenitor cells.

Application

InSolution Purmorphamine, CAS 483367-10-8, is a provided as a 50 mM solution in DMSO. Induces osteoblast differentiation of multipotent mesenchymal progenitor cells C3H10T1/2 & lineage-committed preosteoblasts MC3T3-E1.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/192 µl) solution of Purmorphamine (Cat. No. 540220) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Wu, X., et al. 2004. Chem. Biol.11, 1229.
Wu, X., et al. 2002. J. Am. Chem. Soc.124, 14520.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megumi Kitami et al.
Biochemical and biophysical research communications, 509(1), 222-226 (2018-12-28)
Condylar cartilage is a joint cartilage essential for smooth jaw movement. The importance of ciliary proteins in condylar cartilage development has been reported. However, little is known about how ciliary proteins control the homeostasis of condylar cartilage. Here we show
Arens Taga et al.
Journal of visualized experiments : JoVE, (174) (2021-09-14)
Human pluripotent stem cell-derived astrocytes (hiPSC-A) and neurons (hiPSC-N) provide a powerful tool for modeling Amyotrophic Lateral Sclerosis (ALS) pathophysiology in vitro. Multi-electrode array (MEA) recordings are a means to record electrical field potentials from large populations of neurons and
Aurelie de Rus Jacquet et al.
eLife, 10 (2021-10-01)
Astrocytes are essential cells of the central nervous system, characterized by dynamic relationships with neurons that range from functional metabolic interactions and regulation of neuronal firing activities, to the release of neurotrophic and neuroprotective factors. In Parkinson's disease (PD), dopaminergic
Inge Kinnart et al.
NPJ Parkinson's disease, 10(1), 80-80 (2024-04-10)
The pathogenic effect of SNCA gene multiplications indicates that elevation of wild-type α-synuclein levels is sufficient to cause Parkinson's disease (PD). Mitochondria have been proposed to be a major target of α-synuclein-induced damage. PINK1/parkin/DJ-1-mediated mitophagy is a defense strategy that
Dorien Imberechts et al.
Brain : a journal of neurology, 145(12), 4368-4384 (2022-08-31)
Loss-of-function mutations in the PRKN, PINK1 and PARK7 genes (encoding parkin, PINK1 and DJ-1, respectively) cause autosomal recessive forms of Parkinson's disease. PINK1 and parkin jointly mediate selective autophagy of damaged mitochondria (mitophagy), but the mechanisms by which loss of

Questions

  1. Is this product sterlie?

    1 answer
    1. This product is not sterile.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service