Skip to Content
Merck
All Photos(1)

Key Documents

540025

Millipore

Pluronic® F-127, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered

Non-ionic detergent useful for isolation of membrane proteins.

Synonym(s):

Poloxamer 407, Polyoxypropylenepolyoxyethylene Block Copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
Inquire


Estimated to ship on02 April 2025


Request a Bulk Order

Select a Size

Change View
50 ML
Inquire

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21


Estimated to ship on02 April 2025


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<200 μM aldehyde
<200 μM peroxide
10%±1 detergent

color

clear

shipped in

ambient

storage temp.

2-8°C

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

General description

Conductivity: <50 µmhos.

Application


  • The Influence of Tea Tree Oil on Antifungal Activity and Pharmaceutical Characteristics of Pluronic® F-127 Gel Formulations with Ketoconazole.: This study investigates the effects of tea tree oil on the antifungal properties and pharmaceutical characteristics of Pluronic® F-127 gel formulations containing ketoconazole. The research demonstrated enhanced antifungal activity and improved gel consistency, highlighting the potential of this combination for topical antifungal treatments (Wróblewska et al., 2021).

  • Comprehensive and comparative studies on nanocytotoxicity of glyceryl monooleate- and phytantriol-based lipid liquid crystalline nanoparticles.: This article explores the cytotoxicity of different lipid liquid crystalline nanoparticles, including those stabilized by Pluronic F-127. The findings reveal critical insights into the safety profiles of these nanoparticles for biomedical applications (Jagielski et al., 2021).

  • Co-stabilization of pioglitazone HCL nanoparticles prepared by planetary ball milling: in-vitro and in-vivo evaluation.: This research focuses on the co-stabilization of pioglitazone HCL nanoparticles using Pluronic F-127, demonstrating its effectiveness in enhancing the stability and bioavailability of the drug in both in-vitro and in-vivo settings (Alshora et al., 2020).

  • Dual-responsive (pH/temperature) Pluronic F-127 hydrogel drug delivery system for textile-based transdermal therapy.: This paper presents a novel drug delivery system using Pluronic F-127 hydrogel that responds to both pH and temperature changes. The system is designed for textile-based transdermal therapy, providing controlled drug release (Chatterjee et al., 2019).

  • Formulation and evaluation of novel controlled release of topical pluronic lecithin organogel of mefenamic acid.: This study formulates a controlled-release topical gel using Pluronic F-127 and lecithin to deliver mefenamic acid. The research highlights the gel′s potential for sustained drug release and enhanced therapeutic efficacy (Jhawat et al., 2016).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

10% aqueous solution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany
Pluronic is a registered trademark of BASF

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service