Skip to Content
Merck
All Photos(1)

Key Documents

539844

Sigma-Aldrich

Goat Anti-Rabbit γ-Globulin

lyophilized, Calbiochem®

Synonym(s):

GARGG

Sign Into View Organizational & Contract Pricing

Select a Size

125 U
€125.00

€125.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
125 U
€125.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€125.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

serum

clone

polyclonal

form

lyophilized

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Goat polyclonal antibody supplied as lyophilized, undiluted serum. Recognizes rabbit γ-globulin.
Note: one unit is equivalent to one RIA tube.
One unit of goat anti-rabbit γ-globulin (GARGG) contains enough antibody to maximally precipitate the γ-globulin in 200 µl of 2% normal rabbit serum.
This Goat Anti-Rabbit γ-Globulin is validated for use in Immunoelectrophoresis, Radioimmunoassay for the detection of Rabbit γ-Globulin.

Application

Immunoelectrophoresis (see comments)

Radioimmunoassay (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute with 12.5 ml H₂O. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 weeks at -20°C.

Other Notes

One unit of goat anti-rabbit γ-globulin (GARGG) contains enough antibody to maximally precipitate the γ-globulin in 200 µl of 2% normal rabbit serum. Characteristic precipitin line is observed with normal rabbit serum by immunoelectrophoresis. This antibody has also been reported to work for RIA. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John D Johnson et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 284(2), R422-R432 (2002-10-26)
It has recently been reported that exposure to inescapable tailshock (IS) enhances the release of proinflammatory cytokines following bacterial challenge. However, it is not known whether the level of potentiation of proinflammatory cytokines is sufficient to exaggerate any of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service