Skip to Content
Merck
All Photos(1)

Key Documents

539610

Sigma-Aldrich

PKCζ/ι Pseudosubstrate Inhibitor

The PKCζ/ι Pseudosubstrate Inhibitor controls the biological activity of PKCζ/ι. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKCζ/ι Pseudosubstrate Inhibitor, SIYRRGARRWRKL, Protein Kinase C ζ/ι Pseudosubstrate Inhibitor, SIYRRGARRWRKL, Protein Kinase Cζ/ι Pseudosubstrate Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€205.00

€205.00


Estimated to ship on29 May 2025



Select a Size

Change View
500 μG
€205.00

About This Item

Empirical Formula (Hill Notation):
C76H128N30O16
Molecular Weight:
1718.02
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

glacial acetic acid: 2 mg/mL
water: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A selective, reversible, and substrate competitive inhibitor of protein kinase Cζ and protein kinase Cι (PKCζ/ι) isozymes.
Non-cell-permeable protein kinase Cζ and protein kinase Cι pseudosubstrate sequence peptide that can be used as a selective, reversible, and substrate competitive inhibitor PKCζ/ι in in vitro kinase assays.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PKCζ
Product does not compete with ATP.
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg-Arg-Trp-Arg-Lys-Leu-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sajan, M.P., et al. 1999. J. Biol. Chem.274, 30495.
Laudanna, C., et al. 1998. J. Biol. Chem.273, 30306.
Standaert, M.L., et al. 1997. J. Biol. Chem.272, 30075.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service