Skip to Content
Merck
All Photos(1)

Key Documents

539604

Sigma-Aldrich

PKCη Pseudosubstrate Inhibitor, Myristoylated

The PKCη Pseudosubstrate Inhibitor, Myristoylated controls the biological activity of PKCη. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PKCη Pseudosubstrate Inhibitor, Myristoylated, Myr-TRKRQRAMRRRVHQING-OH, Protein Kinase C η Pseudosubstrate Inhibitor, Myristoylated

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€312.00

€312.00


Estimated to ship on22 May 2025



Select a Size

Change View
500 μG
€312.00

About This Item

Empirical Formula (Hill Notation):
C101H185N41O23S
Molecular Weight:
2373.88
UNSPSC Code:
12352200
NACRES:
NA.77

€312.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible, substrate competitive, and pseudosubstrate inhibitor of the protein kinase Cη isozyme. Useful for studies of PKCη function in intact cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCν
Product competes with ATP.
Reversible: yes

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

Myr-Thr-Arg-Lys-Arg-Gln-Arg-Ala-Met-Arg-Arg-Arg-Val-His-Gln-Ile-Asn-Gly-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Osada, S., et al. 1992. Mol. Cell Biol.12, 3930.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service