539573
PKC Inhibitor Set
The PKC Inhibitor Set controls the biological activity of PKC. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Select a Size
€669.00
Estimated to ship on15 April 2025
Select a Size
About This Item
€669.00
Estimated to ship on15 April 2025
Recommended Products
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
solubility
DMSO: soluble
shipped in
ambient
storage temp.
−20°C
General description
Biochem/physiol Actions
PKC
PKCα, PKCβ
Warning
Reconstitution
Other Notes
Kleinschroth, J., et al. 1993. Bioorg. Med. Chem. Lett.3, 1959.
Martiny-Baron, G.M., et al. 1993. J. Biol. Chem.268, 9194.
Ward, N.E., and O′Brian, C.A. 1993. Biochemistry32, 11903.
Wilkinson, S.E., et al. 1993. Biochem. J.294, 335.
Barg, J., et al. 1992. J. Neurochem.59, 1145.
Gopalakrishna, R., et al. 1992. FEBS Lett.314, 149.
Bruns, R.F., et al. 1991. Biochem. Biophys. Res. Commun.176, 288.
Toullec, D., et al. 1991. J. Biol. Chem.266, 15771.
Herbert, J.M., et al. 1990. Biochem. Biophys. Res. Commun.1172, 993.
Tamaoki, T., et al. 1990. Biotechnology8, 732.
Kobayashi, E., et al. 1989. Biochem. Biophys. Res. Commun.159, 548.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service