Skip to Content
Merck
All Photos(1)

Key Documents

539572

Sigma-Aldrich

Serine/Threonine Kinase Inhibitor Set

The Serine/Threonine Kinase Inhibitor Set controls the biological activity of Serine/Threonine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€599.00

€599.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 SET
€599.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

€599.00


Estimated to ship on29 May 2025


Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: 1 mg/mL (all other inhibitors)
water: 1 mg/mL (Protein Kinase G Inhibitor)

shipped in

ambient

storage temp.

−20°C

General description

A convenient set that offers several highly selective serine/threonine kinase inhibitors. Kinases play an important role in numerous signaling cascades. Highly selective protein kinase inhibitors are useful for dissecting these cascades to determine which specific kinases are involved in producing particular biological effects. The Serine/Threonine Kinase Inhibitor Set contains the following highly selective serine/ threonine kinase inhibitors in one convenient set:



• Bisindolylmaleimide I (Cat. No. 203290), 250 µg

• H-89, Dihydrochloride (Cat. No. 371963), 1 mg

• KN-93 (Cat. No. 422708), 1 mg

• ML-7 (Cat. No. 475880), 1 mg

• Protein Kinase G Inhibitor (Cat. No. 370654), 1 mg

• Staurosporine (Cat. No. 569397), 100 µg

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKA
Secondary Target
PKC, MLCK, Cam kinase II, PKG

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions of each inhibitor are stable for up to 3 months at -20°C.

Other Notes

Martiny, G., et al. 1993. J. Biol. Chem.268, 9194.
Toullec, D., et al. 1991. J. Biol. Chem.266, 15771.
Chijiwa, T., et al. 1990. J. Biol. Chem.265, 5267.
Simu, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Saitoh, M., et al. J. Biol. Chem.262, 7796.
Glass, D.B., et al. 1983. Biochem. J.213, 159.
Seynaeve, C.M., et al. 1994. Mol. Pharmacol.45, 1207.
Meyer, T., et al. 1989. Int. J. Cancer43, 851.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 2

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service