Skip to Content
Merck
All Photos(1)

Key Documents

539515

Sigma-Aldrich

Prothrombin, Human Plasma

Synonym(s):

Prothrombin, Human Plasma, Coagulation Factor II

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€249.00
2 MG
€470.00

€249.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€249.00
2 MG
€470.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€249.00


Estimated to ship on02 June 2025


Request a Bulk Order

description

Merck USA index - 14, 7889

Quality Level

Assay

>95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Native prothrombin from human plasma. Vitamin K-dependent glycoprotein synthesized in the liver. Conversion to thrombin is a key step in the blood coagulation pathway and catalyzes the coagulation of fibrinogen. Clinically, cases of selective deficiency are rare, although, in cases of liver cirrhosis, prothrombin is decreased.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 100 mM NaCl, 20 mM Tris-HCl, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.

Please refer to the label for lot-specific reconstitution volume.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service