Skip to Content
Merck
All Photos(1)

Key Documents

539305

Sigma-Aldrich

Protein G, Alkaline Phosphatase Conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€230.00

€230.00


Estimated to ship on18 May 2025



Select a Size

Change View
500 μG
€230.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

€230.00


Estimated to ship on18 May 2025


form

liquid

Quality Level

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Protein G conjugated to alkaline phosphatase. Does not react with IgA, IgE, or IgM.
Protein G, Alkaline Phosphatase Conjugate is obtained by conjugation of protein G, a cell-surface protein of Streptococcus species to alkaline phosphatase. This conjugate is used as a secondary agent[1] in immunoassay methods to detect polyclonal goat and rabbit antibodies and monoclonal mouse IgG1, IgG2a, and IgG2b.[2]

Application

Protein G, Alkaline Phosphatase Conjugate has been used in:
  • direct enzyme-linked immunosorbent assay (ELISA) (1:1000)[3](1:250)
  • indirect ELISA (1:1000)[4]
  • immunoblot (1:5000)[5]

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM Tris buffer, 1 mM MgCl₂, 0.1 mM ZnCl₂, 1% BSA, 0.1% sodium azide, pH 7.6.

Analysis Note

Single band at 33.8 kDa by SDS-PAGE

Other Notes

p-Nitrophenyl phosphate, 1.0 mg/ml in 10% diethanolamine, pH 9.8, 1 mM MgCl2 at 25°C, was used as substrate to test immunoenzyme reactivity. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sun et al.
Journal of immunological methods, 152(1), 43-48 (1992-07-31)
The IgG binding domains of staphylococcal protein A and streptococcal protein G were expressed as a chimaera using the pGEX vector which has been advocated because its fusion proteins tend to be soluble and easily isolated on immobilised glutathione. This
A O Olarinmoye et al.
Epidemiology and infection, 145(10), 2030-2037 (2017-05-10)
The hamadryas baboon (Papio hamadryas hamadryas) is the only indigenous species of non-human primates (NHP) found in the Kingdom of Saudi Arabia (KSA). There are no peer-reviewed publications on viral infections of the baboons of KSA. Apart from camels, other
B Nilson et al.
Journal of immunoassay, 9(2), 207-225 (1988-01-01)
Protein G, an IgG-binding protein, purified from the surface of group G streptococci, was coupled to alkaline phosphatase. The conjugate was used for detection of polyclonal goat and rabbit antibodies and monoclonal mouse IgG1, IgG2a and IgG2b in an enzyme-linked
Lisa D MacDonald et al.
Human vaccines & immunotherapeutics, 14(1), 59-66 (2017-09-22)
Peptide antigens are combined with an adjuvant in order to increase immunogenicity in vivo. The immunogenicity and safety of a RSV vaccine formulated in a novel oil-based platform, DepoVax™ (DPX), was compared to an alum formulation. A peptide B cell
Anja Reckendorf et al.
International journal for parasitology. Parasites and wildlife, 14, 53-61 (2021-01-26)
Pseudaliid lungworm (Metastrongyloidea) infections and associated secondary bacterial infections may severely affect the health status of harbour porpoises (Phocoena phocoena) in German waters. The presented retrospective analysis including data from 259 harbour porpoises stranded between 2006 and 2018 on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service