Skip to Content
Merck
All Photos(1)

Key Documents

539253

Sigma-Aldrich

Protein A, Peroxidase Conjugate

This Protein A, Peroxidase Conjugate is validated for use in Direct Immunoassay.

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€209.00

€209.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€209.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.54

€209.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

Native protein A from Staphylococcus aureus conjugated to horseradish peroxidase.

Application

Direct Immunoassay (1:10,000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Analysis Note

Single band 50,000 on SDS-PAGE

Other Notes

o-Phenylenediamine, 1.0 mg/ml in citrate buffer, pH 4.5, 0.1% urea hydrogen peroxide at 25°C was used as a substrate for testing conjugate enzyme reactivity. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Tapia-Abellán et al.
Nature chemical biology, 15(6), 560-564 (2019-05-16)
NLRP3 (NOD-like receptor pyrin domain-containing protein 3) is an innate immune sensor that contributes to the development of different diseases, including monogenic autoinflammatory syndromes, gout, atherosclerosis, and Alzheimer's disease. The molecule sulfonylurea MCC950 is a NLRP3 inflammasome inhibitor with potential
Ursula Quitterer et al.
Cell, 176(1-2), 318-333 (2018-12-07)
Preeclampsia is the most frequent pregnancy-related complication worldwide with no cure. While a number of molecular features have emerged, the underlying causal mechanisms behind the disorder remain obscure. Here, we find that increased complex formation between angiotensin II AT1 and
Kwanwoo Lee et al.
The Journal of cell biology, 195(7), 1093-1101 (2011-12-21)
The microtubule-organizing activity of the centrosome oscillates during the cell cycle, reaching its highest level at mitosis. At the onset of mitosis, the centrosome undergoes maturation, which is characterized by a drastic expansion of the pericentriolar matrix (PCM) and a
C F Frey et al.
Parasitology research, 104(6), 1269-1277 (2009-01-09)
Trichinellosis is a zoonotic disease in humans caused by Trichinella spp. According to international regulations and guidelines, serological surveillance can be used to demonstrate the absence of Trichinella spp. in a defined domestic pig population. Most enzyme-linked immunosorbent assay (ELISA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service