Skip to Content
Merck
All Photos(1)

Key Documents

53702

Millipore

Pronase® Protease, Streptomyces griseus

Synonym(s):

Pronase® powder

Sign Into View Organizational & Contract Pricing

Select a Size

10 KU
€91.70
25 KU
€126.00
50 KU
€274.00
250 KU
€873.00
500 KU
€1,440.00

€91.70


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 KU
€91.70
25 KU
€126.00
50 KU
€274.00
250 KU
€873.00
500 KU
€1,440.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€91.70


Estimated to ship on29 May 2025


Request a Bulk Order

description

RTECS - UK9595000

Quality Level

form

lyophilized

specific activity

≥45,000 proteolytic units/g dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A non-specific protease liquefies mucins and digests proteins to free amino acids. Can be used to isolate living chondrocytes. Most active at neutral pH, but it is stable over a wide pH and temperature range. Has an optimal pH of 7.5. Note: 1 KU = 1000 units.
Note: 1 KU = 1000 units.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will liberate a digestion product equivalent to 25 µg of tyrosine per min at 40°C, pH 7.5.

Physical form

Lyophilized from a solution containing calcium acetate as a stabilizer.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for 24 h at 4°C.

Other Notes

Narahashi, Y., et al. 1968. J. Biochem. 64, 427.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PRONASE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole M Templeman et al.
Developmental cell, 54(1), 92-105 (2020-06-17)
Evolutionarily conserved signaling pathways are crucial for adjusting growth, reproduction, and cell maintenance in response to altered environmental conditions or energy balance. However, we have an incomplete understanding of the signaling networks and mechanistic changes that coordinate physiological changes across
T E Davis et al.
Infection and immunity, 15(3), 978-987 (1977-03-01)
Enzymes capable of hydrolyzing cell walls of Blastomyces dermatitidis and chemotypes I and II of Histoplasma capsulatum were prepared in the laboratory or obtained from commercial sources. They included chitinases, beta-1,3-glucanases, beta-1,6-glucanase, and Pronase. Monosaccharides and disaccharides of glucose released
The preparation and characterization of cell walls and the preparation of flagella of Vibrio fetus.
R F Keeler et al.
Journal of general microbiology, 43(3), 439-454 (1966-06-01)
Yu Zhang et al.
Frontiers in molecular neuroscience, 11, 222-222 (2018-07-14)
During development, immature blood vessel networks remodel to form a simplified and efficient vasculature to meet the demand for oxygen and nutrients, and this remodeling process is mainly achieved via the pruning of existing vessels. It has already known that
Rebecca J Casaday et al.
Journal of virology, 78(24), 13501-13511 (2004-11-27)
Capsid assembly among the herpes-group viruses is coordinated by two related scaffolding proteins. In cytomegalovirus (CMV), the main scaffolding constituent is called the assembly protein precursor (pAP). Like its homologs in other herpesviruses, pAP is modified by proteolytic cleavage and

Questions

  1. What is the difference between these two products of yours? first: SKU no. 10165921001 second SKU no. 53702-10KU

    1 answer
    1. The manufacturer/tradename of PRON-RO (10165921001) is Roche where as the manufacturer/tradename of 53702 is Calbiochem®. The SKU and pack sizes are also different for both.
      Answer Source:
      1.https://www.sigmaaldrich.com/IN/en/product/roche/pron-ro
      2. https://www.sigmaaldrich.com/IN/en/product/mm/53702

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service