Skip to Content
Merck
All Photos(1)

Key Documents

5.34359

Sigma-Aldrich

TRPM3 Agonist, CIM0216

Synonym(s):

TRPM3 Agonist, CIM0216, ±-2-(3,4-Dihydroquinolin-1(2H)-yl)-N-(5-methylisoxazol-3-yl)-2-phenylacetamide, Transient Receptor Potential Cation Channel subfamily M member 3 Agonist, CIM-0216

Sign Into View Organizational & Contract Pricing

Select a Size

5343590001
€221.00

€221.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5343590001
€221.00

About This Item

Empirical Formula (Hill Notation):
C21H21N3O2
CAS Number:
Molecular Weight:
347.41
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€221.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to light yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable isoxazolyl-phenylacetamide based compound that acts as a potent, reversible, temperature-dependent and selective activator of both human and murine TRPM3 currents (EC50 = 770 nM for elevating [Ca2+]i levels in HEK293-TRPM3 cells; maximal activation within ~100 s) in a dose-dependent & membrane-delimited manner. Poorly activates human TRPM1, TRPM2, TRPM4, TRPM5, TRPM6, TRPM7, TRPM8 and TRPV1. At higher concentrations, blocks hTRPM2, hTRPM5 & hTRPM8 (by 17%, 34% & 61%, respectively, at 10 µM). Elicits the opening of both the central calcium-conducting pore and the alternative cation permeation pathway. Causes nocifensive behavior in mice (2.5 nmol, intradermal), induces pain, and stimulates the release of CGRP from skin nerve terminals and insulin from pancreatic islets. Shown to display greater efficacy than the canonical TRPM3 agonist, pregnenolone sulfate.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TRPM3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Held, K., et al. 2015. Proc. Natl. Acad. Sci. USA.112, E1363.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service