Skip to Content
Merck
All Photos(2)

Key Documents

5.34356

Sigma-Aldrich

PRMT5 Inhibitor, CMP5

Synonym(s):

PRMT5 Inhibitor, CMP5, 1-(9-Ethyl-9H-carbazol-3-yl)-N-(2-pyridinylmethyl)methanamine, 2HCl, 1-(9-Ethyl-9H-carbazol-3-yl)-N-(pyridin-2-ylmethyl)methanamine, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21N3 · 2HCl
Molecular Weight:
388.33
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 25 mg/mL
water: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable carbazole based compound that selectively inhibits PRMT5 and not PRMT1, PRMT4 & PRMT7 activity. Shown to occupy hPRMT5 catalytic site and block symmetrically dimethylated S2Me-H4R3 & S2Me-H3R8 with no effect on the asymmetric methylation of H4R3 in lymphoblastoid cell lines at 40 µM. Prevents Epstein-Barr virus (EBV)-driven B cell immortalization and restores miR-96 transcriptional activity. Dose-dependently induces lymphoma cell death over normal resting B lymphocytes.
A cell-permeable carbazole based compound that selectively inhibits PRMT5 and not PRMT1, PRMT4 & PRMT7 activity. Shown to occupy hPRMT5 catalytic site and block symmetrically dimethylated S2Me-H4R3 & S2Me-H3R8 with no effect on the asymmetric methylation of H4R3 in lymphoblastoid cell lines at 40 µM. Prevents Epstein-Barr virus (EBV)-driven B cell immortalization and restores miR-96 transcriptional activity. Dose-dependently induces lymphoma cell death over normal resting B lymphocytes.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PRMT5
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as HCl salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Alinari, L., et al. 2015. Blood.125, 2530.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service