Skip to Content
Merck
All Photos(2)

Key Documents

5.33872

Sigma-Aldrich

miR-122 Inhibitor, NSC5476

Synonym(s):

miR-122 Inhibitor, NSC5476, 6-[(4a R,8a S)-(Octahydroquinolin-1(2H)-ylsulfonyl)-1,2,3,4-tetrahydroquinoline, NSC-5476, MLS000736518, microRNA-122 Inhibitor, 6-[(4a R,8a S)-(Octahydro-1(2H)-quinolinylsulfonyl)-1,2,3,4-tetrahydroquinoline, pri-miR-122 Tran, 6-[(4aR,8aS)-(Octahydroquinolin-1(2H)-ylsulfonyl)-1,2,3,4-tetrahydroquinoline, NSC-5476, MLS000736518, microRNA-122 Inhibitor, 6-[(4aR,8aS)-(Octahydro-1(2H)-quinolinylsulfonyl)-1,2,3,4-tetrahydroquinoline, pri-miR-122 Transcrip

Sign Into View Organizational & Contract Pricing

Select a Size

5338720001
€169.00

€169.00


Estimated to ship on22 May 2025



Select a Size

Change View
5338720001
€169.00

About This Item

Empirical Formula (Hill Notation):
C18H26N2O2S
CAS Number:
Molecular Weight:
334.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€169.00


Estimated to ship on22 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N3[C@@H]4[C@@H](CCC3)CCCC4)c1cc2c(cc1)NCCC2

InChI

1S/C18H26N2O2S/c21-23(22,16-9-10-17-15(13-16)6-3-11-19-17)20-12-4-7-14-5-1-2-8-18(14)20/h9-10,13-14,18-19H,1-8,11-12H2/t14-,18+/m1/s1

InChI key

LECPJKATPJRAMB-KDOFPFPSSA-N

General description

A cell-permeable trans-decahydroquinolinylsulfonamide compound that acts as a potent, selective, and reversible inhibitor of miR-122 (EC50 = 600 nM in Huh7 cells transfected with psiCHECK-miR122 reporter plasmid). Induces a 1,250% increase in the relative luciferase signal that is indicative of strong miRNA-122 inhibition. Displays very little activity towards miR-21. Shown to down regulate the levels of mature miR-122 (by 72%) and the primary miR-122 (down to 3%). Shown to reduce hepatitis C virus (HCV) replication in Huh7 cells (47% reduction at 10 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
miR-122
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Young, D.D., et al. 2010. J. Am. Chem. Soc.132, 7976.

Connelly, C.M., et al. 2012. J. Biomol. Screen.17, 822.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service