Skip to Content
Merck
All Photos(1)

Key Documents

5.33717

Sigma-Aldrich

CB-839

≥97% (HPLC), solid, GLS1 inhibitor, Calbiochem®

Synonym(s):

GLS1 Inhibitor III, CB-839, N-(6-(4-(5-((2-Pyridin-2-ylacetyl)amino)-1,3,4-thiadiazol-2-yl)butyl)pyridazin-3-yl)-2-(3-(trifluoromethoxy)phenyl)acetamide, KGA Inhibitor III, 2-(Pyridin-2-yl)-N-(5-(4-(6-(2-(3-(trifluoromethoxy)phenyl)acetamido)pyridazin-3-yl)butyl)-1,3,4-thiadiazol-2

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€238.00

€238.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€238.00

About This Item

Empirical Formula (Hill Notation):
C26H24F3N7O3S
CAS Number:
Molecular Weight:
571.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€238.00


Estimated to ship on22 May 2025


Request a Bulk Order

Product Name

GLS1 Inhibitor III, CB-839,

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to brown

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1cc(ccc1)CC(=O)Nc2nnc(cc2)CCCCc3[s]c(nn3)NC(=O)Cc4ncccc4

InChI

1S/C26H24F3N7O3S/c27-26(28,29)39-20-9-5-6-17(14-20)15-22(37)31-21-12-11-18(33-34-21)7-1-2-10-24-35-36-25(40-24)32-23(38)16-19-8-3-4-13-30-19/h3-6,8-9,11-14H,1-2,7,10,15-16H2,(H,31,34,37)(H,32,36,38)

InChI key

PRAAPINBUWJLGA-UHFFFAOYSA-N

General description

A cell-permeable thiadiazolyl-butyl-pyridazinyl compound that selectively inhibits GLS1 (IC50 = 23 nM & 28 nM, respectively, using murine kidney and brain homogenates), but not GLS2/LGA (up to 5 µM using murine liver homogenates), glutaminase activity in a non-competitive manner, being more potent than the uncompetitive GLS1 inhibitor BPTES, but with much slower inhibition kinetics (IC50/preincubation time ~300 nM/1 min & & 50 nM/1 h for CB-839; 630 nM/1 min & 700 nM/1 h for BPTES; 2 nM rhGAC aa 126-598) & reversibility (Activity recovery t1/2 after free drug removal = 45 min/CB-839 & <3 min/BPTES). CB-839 also displays higher antiproliferation potency than BPTES against triple negative breast cancer (TNBC) cultures (GI50 against MDA-MB-231 in 72 h = 19 nM vs. 2.4 µM with BPTES; GI50 against HCC1806 in 72 h = 55 nM vs. 2.0 µM with BPTES), while neither drug is effective against GLS1-independent growth of estrogen receptor-positive T47D even at 1 µM concentration. Despite a fast clearance in mice (Plasma t1/2<2 h post 50 mg/kg p.o.), good drug exposure is reported in tumor and major organs (>1 nmol/g) 4 h post single 200 mg/10 mL/kg oral dosage among HCC1806 xenograft mice (Drug conc. = 2.1 µM in Plasma & 1.5 nmol/g in 500 mm3 tumor) except brain (~0.2 nmol/g) with most pronounced Glutamine buildup observed in tumor (5-fold) when compared to normal tissues (1- to 2.28-fold of of no treatment controls). Twice daily oral administration (200 mg/kg/12 h) is efficacious in suppressing the expansion of established tumor derived from HIMT-1 (by 54% in 35 d) and patient TNBC (by 59% in 28 d) and complete suppression of JIMT-1 tumor is seen when combined with 5 daily doses of Paclitaxel (10 mg/kg i.v.; Cat. Nos. 580555 & 580556) in the first 5 d of the treatment period.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GLS1
Reversible: yes
Target IC50: 23 nM & 28 nM, respectively, using murine kidney and brain homogenates

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gross, M.I., et al. 2014. Mol. Cancer Ther.13, 890.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service