Skip to Content
Merck

Skip To

5.33162

RAN Translation Inhibitor, NSC311153

Synonym(s):

RAN Translation Inhibitor, NSC311153, 9-Hydroxy-5,11-dimethyl-2-(2-(piperidin-1-yl)ethyl)-6H-pyrido[4,3-b]carbazol-2-ium Bromide, HBr, 9-Hydroxy-5,11-dimethyl-2-(2-(piperidyl)ethyl)-6H-pyrido(4,3-b)carbazolium Bromide, HBr, (GGGGCC)exp RNA Inhibitor, NSC-311153, NSC 311153

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H28BrN3O · xHBr
Molecular Weight:
454.40 (free base basis)
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥96% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥96% (HPLC)

Quality Segment

form

solid

potency

9.7 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

orange

solubility

water: 5 mg/mL, DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable indole fused isoquinolinium salt that binds to the expanded repeat (r(GGGGCC)exp (Kd = 9.7 µM) with modest selectivity. This binding is dependent upon the ionic strength and appears to be significantly weaker in the presence of Na+ and K+. Inhibits repeat-associated non-ATG (RAN) translation and foci Formation in r(GGGGCC)66-expressing HEK293 cells and reduces the levels of poly(GP) and poly(GA) proteins (10% at 25 µM; 47% at to 100 µM). Also shown to inhibit RAN translation and foci formation in (GGGGCC)exp-expressing C9ORF72+ iNeurons (~ 2 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
(r(GGGGCC)exp
Reversible: yes

Packaging

Packaged under inert gas

Physical form

Supplied as HBr salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Su, Z., et al. 2014. Neuron.83, 1043.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters